siRNA Details
| Gene Symbol: Rims3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022931.1 | NP_075220.1 | 8 | 1256 | ||
| GenBank mRNA | AF199334.1 | - | 8 | 1256 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rims3| Gene Symbol: Rims3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022931.1 | NP_075220.1 | 8 | 1256 | ||
| GenBank mRNA | AF199334.1 | - | 8 | 1256 | ||