siRNA Details
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767428.1 | XP_008765650.1 | 6 | 2074 | ||
XM_008767429.1 | XP_008765651.1 | 6 | 1274 | |||
XM_008767430.1 | XP_008765652.1 | 6 | 1305 | |||
XM_008767431.1 | XP_008765653.1 | 6 | 1299 | |||
XM_008767432.1 | XP_008765654.1 | 6 | 1749 | |||
XM_008767433.1 | XP_008765655.1 | 6 | 961 | |||
GenBank mRNA | AB003594.1 | - | 6 | 947 | ||
FQ212368.1 | - | 6 | 1063 | |||
U67137.1 | - | 8 | 2094 | |||
U67987.1 | - | 6 | 888 |
