siRNA Details
| Gene Symbol: Megf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022955.1 | NP_075244.1 | 1925 | |||
| GenBank mRNA | AB011532.1 | - | 1925 | |||
| Gene Symbol: LOC100911486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764417.1 | XP_008762639.1 | 13 | 1586 | ||

Megf6
Genomic Map