siRNA Details
| Gene Symbol: Serpina5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244739.1 | NP_001231668.1 | 2 | 705 | ||
| NM_022957.3 | NP_075246.3 | 2 | 763 | |||
| XM_006240494.2 | XP_006240556.1 | 5 | 2878 | |||
| GenBank mRNA | AB013128.1 | - | 2 | 636 | ||
| BC081797.1 | - | 2 | 673 | 7112069 | ||
| BC097315.1 | - | 2 | 720 | 7453034 | ||

Serpina5
Genomic Map