siRNA Details
| Gene Symbol: Pik3c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022958.2 | NP_075247.1 | 15, 16 | 1729 | ||
| XM_006254488.2 | XP_006254550.1 | 15, 16 | 1807 | |||
| GenBank mRNA | AJ006710.1 | - | 15, 16 | 1711 | ||
| BC061981.1 | - | 15, 16 | 1729 | 5597998 | ||

Pik3c3
Genomic Map