siRNA Details
| Gene Symbol: Alas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024484.2 | NP_077810.2 | 4 | 643 | ||
| GenBank mRNA | BC061793.1 | - | 4 | 643 | 5597891 | |
| J03190.1 | - | 3 | 567 | |||
| J04044.1 | - | 3 | 574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alas1| Gene Symbol: Alas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024484.2 | NP_077810.2 | 4 | 643 | ||
| GenBank mRNA | BC061793.1 | - | 4 | 643 | 5597891 | |
| J03190.1 | - | 3 | 567 | |||
| J04044.1 | - | 3 | 574 | |||