siRNA Details
Gene Symbol: Amotl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031717.1 | NP_113905.1 | 2 | 622 | ||
XM_006243680.2 | XP_006243742.1 | 2 | 724 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Amotl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031717.1 | NP_113905.1 | 2 | 622 | ||
XM_006243680.2 | XP_006243742.1 | 2 | 724 |