siRNA Details
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031719.1 | NP_113907.1 | 3, 4 | 408 | ||
XM_006229792.2 | XP_006229854.1 | 3, 4 | 471 | |||
GenBank mRNA | D13985.1 | - | 3, 4 | 478 | ||
D17698.1 | - | 3, 4 | 391 | |||
FQ212666.1 | - | 1, 2 | 90 | |||
FQ232880.1 | - | 1, 2 | 90 | |||
L26450.1 | - | 3, 4 | 408 |
