siRNA Details
| Gene Symbol: Limk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031727.2 | NP_113915.2 | 7, 8 | 1066 | ||
| XM_006249185.2 | XP_006249247.1 | 6, 7 | 4158 | |||
| GenBank mRNA | AW529884.1 | - | 1, 2 | 125 | ||
| D31873.1 | - | 7, 8 | 1091 | |||

Limk1
Genomic Map