siRNA Details
| Gene Symbol: Snap91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031728.1 | NP_113916.1 | 22, 23 | 2111 | ||
| XM_006243491.2 | XP_006243553.1 | 22, 23 | 2200 | |||
| XM_006243492.2 | XP_006243554.1 | 22, 23 | 2261 | |||
| XM_006243493.2 | XP_006243555.1 | 21, 22 | 2194 | |||
| XM_006243494.2 | XP_006243556.1 | 22, 23 | 2200 | |||
| XM_006243495.2 | XP_006243557.1 | 22, 23 | 2200 | |||
| XM_006243497.2 | XP_006243559.1 | 21, 22 | 2092 | |||
| XM_008766455.1 | XP_008764677.1 | 22, 23 | 2156 | |||
| XM_008766456.1 | XP_008764678.1 | 21, 22 | 2185 | |||
| GenBank mRNA | X68877.1 | - | 22, 23 | 2054 | ||
| X68878.1 | - | 22, 23 | 2111 | |||

Snap91
Genomic Map