siRNA Details
| Gene Symbol: Gabrq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031733.1 | NP_113921.1 | 9 | 1228 | ||
| XM_006229516.2 | XP_006229578.1 | 10 | 1392 | |||
| GenBank mRNA | AF419333.1 | - | 9 | 1228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrq| Gene Symbol: Gabrq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031733.1 | NP_113921.1 | 9 | 1228 | ||
| XM_006229516.2 | XP_006229578.1 | 10 | 1392 | |||
| GenBank mRNA | AF419333.1 | - | 9 | 1228 | ||