siRNA Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031741.1 | NP_113929.1 | 11 | 1327 | ||
| GenBank mRNA | D13871.1 | - | 11 | 1327 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a5| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031741.1 | NP_113929.1 | 11 | 1327 | ||
| GenBank mRNA | D13871.1 | - | 11 | 1327 | ||