siRNA Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031741.1 | NP_113929.1 | 2, 3 | 173 | ||
| GenBank mRNA | BC076378.1 | - | 2, 3 | 158 | ||
| BC161843.1 | - | 2, 3 | 160 | 7127817 | ||
| D13871.1 | - | 2, 3 | 173 | |||
| D28562.1 | - | 2, 3 | 132 | |||
| L05195.1 | - | 2, 3 | 145 | |||

Slc2a5
Genomic Map