siRNA Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032462.2 | NP_115851.1 | 2 | 153 | ||
| XM_006234989.2 | XP_006235051.1 | 2 | 286 | |||
| GenBank mRNA | AB043892.1 | - | 1 | 132 | ||
| AF297118.1 | - | 2 | 155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnip3| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032462.2 | NP_115851.1 | 2 | 153 | ||
| XM_006234989.2 | XP_006235051.1 | 2 | 286 | |||
| GenBank mRNA | AB043892.1 | - | 1 | 132 | ||
| AF297118.1 | - | 2 | 155 | |||