siRNA Details
| Gene Symbol: Slc13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031746.1 | NP_113934.1 | 10 | 1461 | ||
| XM_006246947.2 | XP_006247009.1 | 10 | 1453 | |||
| GenBank mRNA | AB001321.1 | - | 10 | 1464 | ||
| AF058714.1 | - | 10 | 1461 | |||
| U51153.1 | - | 10 | 1453 | |||

Slc13a2
Genomic Map