siRNA Details
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023023.1 | NP_075412.1 | 2 | 141 | ||
| GenBank mRNA | AB029432.1 | - | 1 | 18 | ||
| AJ131436.1 | - | 2 | 141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpysl5| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023023.1 | NP_075412.1 | 2 | 141 | ||
| GenBank mRNA | AB029432.1 | - | 1 | 18 | ||
| AJ131436.1 | - | 2 | 141 | |||