siRNA Details
| Gene Symbol: Dync2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023024.1 | NP_075413.1 | 47 | 8032 | ||
| GenBank mRNA | AB041881.1 | - | 47 | 8032 | ||
| Gene Symbol: LOC103689995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765895.1 | XP_008764117.1 | 1095 | |||
| XM_008765896.1 | XP_008764118.1 | 1074 | ||||

Dync2h1
Genomic Map