siRNA Details
Gene Symbol: Prkaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019142.2 | NP_062015.2 | 7 | 1088 | ||
GenBank mRNA | U40819.1 | - | 7 | 1055 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prkaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019142.2 | NP_062015.2 | 7 | 1088 | ||
GenBank mRNA | U40819.1 | - | 7 | 1055 |