siRNA Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023093.1 | NP_075581.1 | 6, 7 | 849 | ||
| GenBank mRNA | BC061830.1 | - | 6, 7 | 849 | 5599635 | |
| FQ214146.1 | - | 5, 6 | 691 | |||
| FQ225416.1 | - | 1, 2 | 164 | |||
| FQ225735.1 | - | 1, 2 | 164 | |||
| FQ227531.1 | - | 1, 2 | 164 | |||
| J05266.1 | - | 6, 7 | 770 | |||

Atp5a1
Genomic Map