siRNA Details
Gene Symbol: Mgat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023095.1 | NP_075583.1 | 10 | 1388 | ||
XM_008769499.1 | XP_008767721.1 | 6 | 932 | |||
GenBank mRNA | L14284.1 | - | 10 | 1388 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023095.1 | NP_075583.1 | 10 | 1388 | ||
XM_008769499.1 | XP_008767721.1 | 6 | 932 | |||
GenBank mRNA | L14284.1 | - | 10 | 1388 |