siRNA Details
| Gene Symbol: Nmur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023100.1 | NP_075588.1 | 1 | 813 | ||
| GenBank mRNA | AB038649.1 | - | 1 | 813 | ||
| AF242873.1 | - | 1 | 783 | |||
| BC081796.1 | - | 840 | ||||
More Information
Set Membership: |
|

Nmur1
Genomic Map