siRNA Details
| Gene Symbol: Aacs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023104.1 | NP_075592.1 | 2 | 305 | ||
| GenBank mRNA | AB026291.1 | - | 2 | 305 | ||
| BC061803.1 | - | 2 | 346 | 5598532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aacs| Gene Symbol: Aacs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023104.1 | NP_075592.1 | 2 | 305 | ||
| GenBank mRNA | AB026291.1 | - | 2 | 305 | ||
| BC061803.1 | - | 2 | 346 | 5598532 | ||