siRNA Details
| Gene Symbol: Arhgef9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023957.1 | NP_076447.1 | 9 | 1733 | ||
| GenBank mRNA | AJ250425.1 | - | 9 | 1733 | ||
| AJ302676.1 | - | 8 | 1553 | |||
| Gene Symbol: LOC100912165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773295.1 | XP_008771517.1 | 2 | 469 | ||
| XM_008773296.1 | XP_008771518.1 | 472 | ||||

Arhgef9
Genomic Map