siRNA Details
| Gene Symbol: Adamts4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023959.1 | NP_076449.1 | 1 | 318 | ||
| GenBank mRNA | CB614460.1 | - | 1 | 105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts4| Gene Symbol: Adamts4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023959.1 | NP_076449.1 | 1 | 318 | ||
| GenBank mRNA | CB614460.1 | - | 1 | 105 | ||