siRNA Details
| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023962.2 | NP_076452.1 | 5 | 756 | ||
| GenBank mRNA | AB052170.2 | - | 5 | 756 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdgfd| Gene Symbol: Pdgfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023962.2 | NP_076452.1 | 5 | 756 | ||
| GenBank mRNA | AB052170.2 | - | 5 | 756 | ||