siRNA Details
| Gene Symbol: Cybb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023965.1 | NP_076455.1 | 13 | 1682 | ||
| GenBank mRNA | AF298656.3 | - | 13 | 1682 | ||
| AJ295950.1 | - | 13 | 1682 | |||
| FQ217794.1 | - | 1751 | ||||
| FQ231233.1 | - | 1734 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cybb| Gene Symbol: Cybb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023965.1 | NP_076455.1 | 13 | 1682 | ||
| GenBank mRNA | AF298656.3 | - | 13 | 1682 | ||
| AJ295950.1 | - | 13 | 1682 | |||
| FQ217794.1 | - | 1751 | ||||
| FQ231233.1 | - | 1734 | ||||