siRNA Details
| Gene Symbol: Cybb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023965.1 | NP_076455.1 | 9 | 957 | ||
| GenBank mRNA | AJ295950.1 | - | 9 | 957 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cybb| Gene Symbol: Cybb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023965.1 | NP_076455.1 | 9 | 957 | ||
| GenBank mRNA | AJ295950.1 | - | 9 | 957 | ||