siRNA Details
Gene Symbol: Trpv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023970.1 | NP_076460.1 | 7 | 1304 | ||
XM_006249466.2 | XP_006249528.1 | 7 | 1333 | |||
GenBank mRNA | AF263521.1 | - | 7 | 1304 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trpv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023970.1 | NP_076460.1 | 7 | 1304 | ||
XM_006249466.2 | XP_006249528.1 | 7 | 1333 | |||
GenBank mRNA | AF263521.1 | - | 7 | 1304 |