siRNA Details
| Gene Symbol: Wdr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023975.1 | NP_076465.1 | 10 | 1165 | ||
| XM_008772171.1 | XP_008770393.1 | 11 | 1372 | |||
| GenBank mRNA | AF192379.1 | - | 10 | 1165 | ||
| AF305813.1 | - | 10 | 1165 | |||

Wdr7
Genomic Map