siRNA Details
Gene Symbol: Shank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031751.2 | NP_113939.2 | 8 | 1147 | ||
XM_008759414.1 | XP_008757636.1 | 7 | 1214 | |||
XM_008759415.1 | XP_008757637.1 | 7 | 1214 | |||
XM_008759416.1 | XP_008757638.1 | 7 | 1214 | |||
XM_008759417.1 | XP_008757639.1 | 7 | 1214 | |||
GenBank mRNA | AF102855.2 | - | 7 | 1059 | ||
AF131951.1 | - | 7 | 1068 | |||
AF159046.1 | - | 8 | 1147 |
