siRNA Details
| Gene Symbol: Akr1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031000.3 | NP_112262.1 | 9 | 1003 | ||
| GenBank mRNA | BC059133.1 | - | 9 | 1282 | 6918695 | |
| D10854.1 | - | 9 | 1003 | |||
| FQ212779.1 | - | 9 | 1278 | |||
| FQ213220.1 | - | 8 | 950 | |||
| FQ213467.1 | - | 9 | 1278 | |||
| FQ213483.1 | - | 9 | 1274 | |||
| FQ213491.1 | - | 9 | 1276 | |||
| FQ213579.1 | - | 9 | 1246 | |||
| FQ213776.1 | - | 9 | 1245 | |||
| FQ214173.1 | - | 9 | 1246 | |||
| FQ214441.1 | - | 9 | 1022 | |||
| FQ214642.1 | - | 1259 | ||||
| FQ214700.1 | - | 9 | 1219 | |||
| FQ215620.1 | - | 9 | 1070 | |||
| FQ215772.1 | - | 9 | 1246 | |||
| FQ218383.1 | - | 9 | 1246 | |||
| FQ218721.1 | - | 9 | 1245 | |||
| FQ218729.1 | - | 9 | 1245 | |||
| FQ220055.1 | - | 7 | 846 | |||
| FQ226221.1 | - | 9 | 1278 | |||
| FQ228131.1 | - | 9 | 1279 | |||
| FQ228132.1 | - | 9 | 1278 | |||
| FQ228842.1 | - | 9 | 1274 | |||
| FQ233156.1 | - | 9 | 1103 | |||

Akr1a1
Genomic Map