siRNA Details
| Gene Symbol: Golph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023977.2 | NP_076467.2 | 4 | 991 | ||
| GenBank mRNA | AF311954.1 | - | 4 | 882 | ||
| BC092568.1 | - | 4 | 991 | 7307277 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Golph3| Gene Symbol: Golph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023977.2 | NP_076467.2 | 4 | 991 | ||
| GenBank mRNA | AF311954.1 | - | 4 | 882 | ||
| BC092568.1 | - | 4 | 991 | 7307277 | ||