siRNA Details
| Gene Symbol: Mcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034009.1 | NP_001029181.1 | 5 | 536 | ||
| NM_023983.3 | NP_076473.2 | 5 | 536 | |||
| GenBank mRNA | AB035506.1 | - | 5 | 534 | ||
| AB035507.1 | - | 5 | 534 | |||
| BC070916.1 | - | 5 | 536 | 7105099 | ||

Mcam
Genomic Map