siRNA Details
| Gene Symbol: Trib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023985.1 | NP_076475.1 | 3 | 1288 | ||
| GenBank mRNA | AF205438.1 | - | 3 | 664 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trib1| Gene Symbol: Trib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023985.1 | NP_076475.1 | 3 | 1288 | ||
| GenBank mRNA | AF205438.1 | - | 3 | 664 | ||