siRNA Details
| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024002.1 | NP_076492.1 | 14 | 2005 | ||
| XM_008771506.1 | XP_008769728.1 | 14 | 2055 | |||
| GenBank mRNA | AJ251245.1 | - | 14 | 2005 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Secisbp2| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024002.1 | NP_076492.1 | 14 | 2005 | ||
| XM_008771506.1 | XP_008769728.1 | 14 | 2055 | |||
| GenBank mRNA | AJ251245.1 | - | 14 | 2005 | ||