siRNA Details
| Gene Symbol: Duox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024141.1 | NP_077055.1 | 17 | 2303 | ||
| XM_006234862.2 | XP_006234924.1 | 19 | 3267 | |||
| GenBank mRNA | AF237962.1 | - | 17 | 2303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Duox2| Gene Symbol: Duox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024141.1 | NP_077055.1 | 17 | 2303 | ||
| XM_006234862.2 | XP_006234924.1 | 19 | 3267 | |||
| GenBank mRNA | AF237962.1 | - | 17 | 2303 | ||