siRNA Details
| Gene Symbol: Duox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024141.1 | NP_077055.1 | 26 | 3580 | ||
| XM_006234862.2 | XP_006234924.1 | 28 | 4544 | |||
| GenBank mRNA | AF237962.1 | - | 26 | 3580 | ||
| AF547268.1 | - | 4 | 589 | |||

Duox2
Genomic Map