siRNA Details
| Gene Symbol: Slc27a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024143.2 | NP_077057.1 | 2 | 888 | ||
| GenBank mRNA | AF242189.1 | - | 2 | 889 | ||
| BC091147.1 | - | 2 | 911 | 7370589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc27a5| Gene Symbol: Slc27a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024143.2 | NP_077057.1 | 2 | 888 | ||
| GenBank mRNA | AF242189.1 | - | 2 | 889 | ||
| BC091147.1 | - | 2 | 911 | 7370589 | ||