siRNA Details
| Gene Symbol: Dck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024158.1 | NP_077072.1 | 4, 5 | 577 | ||
| GenBank mRNA | FQ212366.1 | - | 882 | |||
| L33894.1 | - | 4, 5 | 577 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dck| Gene Symbol: Dck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024158.1 | NP_077072.1 | 4, 5 | 577 | ||
| GenBank mRNA | FQ212366.1 | - | 882 | |||
| L33894.1 | - | 4, 5 | 577 | |||