siRNA Details
| Gene Symbol: Fstl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024369.2 | NP_077345.1 | 9, 10 | 877 | ||
| GenBank mRNA | AY325203.1 | - | 1255 | |||
| BC087014.1 | - | 9, 10 | 877 | 7189359 | ||
| U06864.1 | - | 9, 10 | 852 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fstl1| Gene Symbol: Fstl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024369.2 | NP_077345.1 | 9, 10 | 877 | ||
| GenBank mRNA | AY325203.1 | - | 1255 | |||
| BC087014.1 | - | 9, 10 | 877 | 7189359 | ||
| U06864.1 | - | 9, 10 | 852 | |||