siRNA Details
Gene Symbol: Gabrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024370.3 | NP_077346.3 | 5 | 630 | ||
GenBank mRNA | M81142.1 | - | 5 | 612 | ||
X63324.1 | - | 686 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gabrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024370.3 | NP_077346.3 | 5 | 630 | ||
GenBank mRNA | M81142.1 | - | 5 | 612 | ||
X63324.1 | - | 686 |