siRNA Details
Gene Symbol: Slc6a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024372.2 | NP_077348.1 | 8 | 1004 | ||
GenBank mRNA | M95738.1 | - | 8 | 1007 | ||
M95763.1 | - | 8 | 1004 | |||
S42358.1 | - | 8 | 1007 |
