siRNA Details
| Gene Symbol: Slc6a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024372.2 | NP_077348.1 | 14 | 1817 | ||
| GenBank mRNA | M95738.1 | - | 14 | 1820 | ||
| M95763.1 | - | 14 | 1817 | |||
| S42358.1 | - | 14 | 1820 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a11| Gene Symbol: Slc6a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024372.2 | NP_077348.1 | 14 | 1817 | ||
| GenBank mRNA | M95738.1 | - | 14 | 1820 | ||
| M95763.1 | - | 14 | 1817 | |||
| S42358.1 | - | 14 | 1820 | |||