siRNA Details
| Gene Symbol: Grid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024378.1 | NP_077354.1 | 15 | 2604 | ||
| GenBank mRNA | U08255.1 | - | 15 | 2604 | ||
| Z17238.1 | - | 13 | 2237 | |||
| Gene Symbol: LOC103690098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252770.2 | XP_006252832.1 | 4272 | |||

Grid1
Genomic Map