siRNA Details
| Gene Symbol: Grid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024378.1 | NP_077354.1 | 10 | 1546 | ||
| GenBank mRNA | U08255.1 | - | 10 | 1546 | ||
| Z17238.1 | - | 8 | 1179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grid1| Gene Symbol: Grid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024378.1 | NP_077354.1 | 10 | 1546 | ||
| GenBank mRNA | U08255.1 | - | 10 | 1546 | ||
| Z17238.1 | - | 8 | 1179 | |||