siRNA Details
| Gene Symbol: Nr4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024388.2 | NP_077364.2 | 2 | 936 | ||
| XM_006242356.2 | XP_006242418.1 | 2 | 3763 | |||
| XM_006242358.2 | XP_006242420.1 | 2 | 1691 | |||
| GenBank mRNA | BC097313.1 | - | 2 | 929 | 7458540 | |
| U17254.1 | - | 2 | 929 | |||

Nr4a1
Genomic Map