siRNA Details
| Gene Symbol: Hpgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024390.2 | NP_077366.2 | 1, 2 | 114 | ||
| GenBank mRNA | BC062399.1 | - | 1, 2 | 114 | 5599288 | |
| U44750.1 | - | 1, 2 | 130 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpgd| Gene Symbol: Hpgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024390.2 | NP_077366.2 | 1, 2 | 114 | ||
| GenBank mRNA | BC062399.1 | - | 1, 2 | 114 | 5599288 | |
| U44750.1 | - | 1, 2 | 130 | |||