siRNA Details
| Gene Symbol: Hsd17b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024391.1 | NP_077367.1 | 5 | 1175 | ||
| GenBank mRNA | BC088134.1 | - | 5 | 1166 | 7326035 | |
| X91234.1 | - | 5 | 1175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd17b2| Gene Symbol: Hsd17b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024391.1 | NP_077367.1 | 5 | 1175 | ||
| GenBank mRNA | BC088134.1 | - | 5 | 1166 | 7326035 | |
| X91234.1 | - | 5 | 1175 | |||