siRNA Details
Gene Symbol: Hsd17b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024391.1 | NP_077367.1 | 3 | 621 | ||
GenBank mRNA | BC088134.1 | - | 3 | 612 | 7326035 | |
X91234.1 | - | 3 | 621 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hsd17b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024391.1 | NP_077367.1 | 3 | 621 | ||
GenBank mRNA | BC088134.1 | - | 3 | 612 | 7326035 | |
X91234.1 | - | 3 | 621 |