siRNA Details
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024394.2 | NP_077370.2 | 9 | 1542 | ||
XM_006243021.2 | XP_006243083.1 | 9 | 1571 | |||
GenBank mRNA | D49395.1 | - | 9 | 1433 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024394.2 | NP_077370.2 | 9 | 1542 | ||
XM_006243021.2 | XP_006243083.1 | 9 | 1571 | |||
GenBank mRNA | D49395.1 | - | 9 | 1433 |